The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005068.2 | NP_005059.2 | 7 - 8 | 1060 | 87 |
XM_005267100.2 | XP_005267157.1 | 8 - 9 | 1636 | 87 | |
XM_011536072.2 | XP_011534374.1 | 8 - 9 | 1636 | 87 | |
XM_017011197.1 | XP_016866686.1 | 8 - 9 | 1368 | 87 | |
GenBank mRNA | AK308090.1 | - | 8 - 9 | 1636 | 87 |
U70212.1 | - | 7 - 8 | 1069 | 87 |