The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126054.2 | NP_001119526.1 | 1 - 2 | 107 | 70 |
NM_001126055.2 | NP_001119527.1 | 1 - 2 | 107 | 70 | |
NM_003688.3 | NP_003679.2 | 1 - 2 | 107 | 70 | |
XM_005272686.4 | XP_005272743.1 | 1 - 2 | 598 | 70 | |
XM_006724566.3 | XP_006724629.1 | 1 - 2 | 598 | 70 | |
XM_011543993.2 | XP_011542295.1 | 1 - 2 | 159 | 70 | |
XM_011543994.2 | XP_011542296.1 | 1 - 2 | 159 | 70 | |
XM_011543995.2 | XP_011542297.1 | 1 - 2 | 159 | 70 | |
XM_011543996.2 | XP_011542298.1 | 1 - 2 | 159 | 70 | |
GenBank mRNA | AB208859.1 | - | 1 - 2 | 51 | 70 |
AF032119.1 | - | 1 - 2 | 88 | 70 | |
AF035582.1 | - | 1 - 2 | 76 | 70 | |
AK313399.1 | - | 1 - 2 | 260 | 70 | |
AY705392.1 | - | 1 - 2 | 67 | 70 | |
BC117311.1 | - | 1 - 2 | 80 | 70 | |
BC143454.1 | - | 1 - 2 | 80 | 70 | |
BC143456.1 | - | 1 - 2 | 80 | 70 | |
DN993605.1 | - | 1 - 2 | 115 | 70 |