The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126054.2 | NP_001119526.1 | 2 - 3 | 221 | 78 |
NM_001126055.2 | NP_001119527.1 | 2 - 3 | 221 | 78 | |
NM_003688.3 | NP_003679.2 | 2 - 3 | 221 | 78 | |
XM_005272686.4 | XP_005272743.1 | 2 - 3 | 712 | 78 | |
XM_006724566.3 | XP_006724629.1 | 2 - 3 | 712 | 78 | |
GenBank mRNA | AB208859.1 | - | 2 - 3 | 165 | 78 |
AF032119.1 | - | 2 - 3 | 202 | 78 | |
AF035582.1 | - | 2 - 3 | 190 | 78 | |
AF262405.1 | - | 1 - 2 | 97 | 78 | |
AK313399.1 | - | 2 - 3 | 374 | 78 | |
AY705392.1 | - | 2 - 3 | 181 | 78 | |
BC117311.1 | - | 2 - 3 | 194 | 78 | |
BC143454.1 | - | 2 - 3 | 194 | 78 | |
BC143456.1 | - | 2 - 3 | 194 | 78 | |
DN993605.1 | - | 2 - 3 | 229 | 78 |