Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPH3A
Entrez Gene ID
22895
Gene Name
rabphilin 3A
Gene Aliases
-
Chromosome Location
Chr.12: 112791538 - 112898881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143854.1 NP_001137326.1 19 - 20 2189 62
NM_014954.3 NP_055769.2 18 - 19 2177 62
XM_011538048.1 XP_011536350.1 19 - 20 1993 62
XM_017019034.1 XP_016874523.1 11 - 12 1447 62
GenBank mRNA AB023202.1 - 19 - 20 2110 62
AK295696.1 - 19 - 20 2143 62
AK296296.1 - 17 - 18 1728 62
AK316112.1 - 19 - 20 2024 62
BC002605.2 - 17 - 18 1912 62
BC009843.2 - 17 - 18 1838 62
BC017259.2 - 18 - 19 1984 62
BX647281.1 - 11 - 12 2783 62

View Full Product Details