Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPH3A
Entrez Gene ID
22895
Gene Name
rabphilin 3A
Gene Aliases
-
Chromosome Location
Chr.12: 112791538 - 112898881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143854.1 NP_001137326.1 1 - 2 267 84
NM_014954.3 NP_055769.2 1 - 2 267 84
GenBank mRNA AB023202.1 - 1 - 2 188 84
AK316112.1 - 1 - 2 102 84
BC017259.2 - 1 - 2 74 84
DA332963.1 - 1 - 2 267 84

View Full Product Details