The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135648.2 | NP_001129120.1 | 27 - 28 | 4180 | 74 |
NM_001291981.1 | NP_001278910.1 | 29 - 30 | 4228 | 74 | |
NM_001291984.1 | NP_001278913.1 | 26 - 27 | 4159 | 74 | |
NM_002844.3 | NP_002835.2 | 26 - 27 | 4162 | 74 | |
XM_006715537.2 | XP_006715600.1 | 27 - 28 | 4228 | 74 | |
XM_011536014.2 | XP_011534316.1 | 29 - 30 | 4276 | 74 | |
XM_011536015.2 | XP_011534317.1 | 28 - 29 | 4267 | 74 | |
XM_011536016.2 | XP_011534318.1 | 28 - 29 | 3903 | 74 | |
XM_011536017.2 | XP_011534319.1 | 30 - 31 | 4519 | 74 | |
XM_011536018.2 | XP_011534320.1 | 30 - 31 | 4192 | 74 | |
XM_017011145.1 | XP_016866634.1 | 28 - 29 | 4195 | 74 | |
XM_017011146.1 | XP_016866635.1 | 28 - 29 | 4144 | 74 | |
XM_017011147.1 | XP_016866636.1 | 28 - 29 | 4122 | 74 | |
XM_017011148.1 | XP_016866637.1 | 27 - 28 | 4101 | 74 | |
XM_017011149.1 | XP_016866638.1 | 17 - 18 | 1780 | 74 | |
XM_017011150.1 | XP_016866639.1 | 16 - 17 | 1767 | 74 | |
GenBank mRNA | AF533875.1 | - | 26 - 27 | 3927 | 74 |
BC140775.1 | - | 27 - 28 | 3982 | 74 | |
BC144512.1 | - | 27 - 28 | 3982 | 74 | |
BC144513.1 | - | 29 - 30 | 4030 | 74 | |
BX647498.1 | - | 4 - 5 | 408 | 74 | |
CR749277.1 | - | 26 - 27 | 4103 | 74 | |
L77886.1 | - | 26 - 27 | 3861 | 74 | |
Z70660.1 | - | 26 - 27 | 3843 | 74 |