The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014797.2 | NP_055612.2 | 4 - 5 | 1374 | 63 |
GenBank mRNA | AB007901.1 | - | 4 - 5 | 1374 | 63 |
AK309697.1 | - | 4 - 5 | 1328 | 63 | |
BC117374.1 | - | 3 - 4 | 1242 | 63 | |
BC117376.1 | - | 3 - 4 | 1242 | 63 | |
BX648883.1 | - | - | 550 | 63 |