Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIN2
Entrez Gene ID
54453
Gene Name
Ras and Rab interactor 2
Gene Aliases
MACS, RASSF4
Chromosome Location
Chr.20: 19757610 - 20002459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005260733.2 XP_005260790.1 3 - 4 376 77

View Full Product Details