The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129.4 | NP_001120.3 | 11 - 12 | 1705 | 64 |
XM_011515162.1 | XP_011513464.1 | 10 - 11 | 1668 | 64 | |
GenBank mRNA | AB209744.1 | - | 11 - 12 | 1659 | 64 |
AF053944.1 | - | 11 - 12 | 1539 | 64 | |
BC038588.1 | - | 11 - 12 | 1705 | 64 | |
D86479.1 | - | 5 - 6 | 461 | 64 |