Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF32
Entrez Gene ID
140545
Gene Name
ring finger protein 32
Gene Aliases
FKSG33, HSD15, LMBR2
Chromosome Location
Chr.7: 156640286 - 156678333 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005249522.4 XP_005249579.1 1 - 2 64 102

View Full Product Details