Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLG1
Entrez Gene ID
1739
Gene Name
discs large MAGUK scaffold protein 1
Gene Aliases
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
Chromosome Location
Chr.3: 197042560 - 197299272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098424.1 NP_001091894.1 14 - 15 1727 72
NM_001204386.1 NP_001191315.1 13 - 14 1628 72
NM_001204387.1 NP_001191316.1 9 - 10 1512 72
NM_001204388.1 NP_001191317.1 9 - 10 1512 72
NM_001290983.1 NP_001277912.1 14 - 15 1796 72
NM_004087.2 NP_004078.2 14 - 15 1727 72
XM_005269289.3 XP_005269346.1 14 - 15 2009 72
XM_011512502.2 XP_011510804.1 14 - 15 2009 72
XM_011512503.1 XP_011510805.1 13 - 14 1907 72
XM_011512505.1 XP_011510807.1 13 - 14 1907 72
XM_011512506.1 XP_011510808.1 12 - 13 1853 72
XM_011512509.1 XP_011510811.1 13 - 14 1907 72
XM_017005800.1 XP_016861289.1 14 - 15 2144 72
XM_017005801.1 XP_016861290.1 14 - 15 1664 72
XM_017005802.1 XP_016861291.1 14 - 15 1734 72
XM_017005803.1 XP_016861292.1 14 - 15 1655 72
XM_017005804.1 XP_016861293.1 14 - 15 2009 72
XM_017005805.1 XP_016861294.1 14 - 15 1826 72
XM_017005806.1 XP_016861295.1 13 - 14 1907 72
XM_017005807.1 XP_016861296.1 13 - 14 2006 72
XM_017005808.1 XP_016861297.1 13 - 14 1565 72
XM_017005809.1 XP_016861298.1 13 - 14 1667 72
XM_017005810.1 XP_016861299.1 13 - 14 1565 72
XM_017005811.1 XP_016861300.1 12 - 13 1853 72
XM_017005812.1 XP_016861301.1 12 - 13 1507 72
XM_017005813.1 XP_016861302.1 12 - 13 1511 72
XM_017005814.1 XP_016861303.1 12 - 13 1614 72
XM_017005815.1 XP_016861304.1 12 - 13 1853 72
XM_017005816.1 XP_016861305.1 13 - 14 2006 72
XM_017005817.1 XP_016861306.1 13 - 14 1537 72
XM_017005818.1 XP_016861307.1 13 - 14 1639 72
XM_017005819.1 XP_016861308.1 13 - 14 1556 72
XM_017005820.1 XP_016861309.1 12 - 13 1571 72
XM_017005821.1 XP_016861310.1 10 - 11 4012 72
XM_017005822.1 XP_016861311.1 10 - 11 4014 72
XM_017005823.1 XP_016861312.1 9 - 10 3909 72
XM_017005824.1 XP_016861313.1 4 - 5 528 72
GenBank mRNA AB209536.2 - 6 - 7 850 72
AK293967.1 - 9 - 10 1231 72
AK294772.1 - 9 - 10 1239 72
AK294855.1 - 9 - 10 1512 72
AK310596.1 - 13 - 14 1566 72
AK310893.1 - 14 - 15 2011 72
BC140841.1 - 14 - 15 1692 72
BC144651.1 - 13 - 14 1593 72
EF553524.1 - 14 - 15 1796 72
U13896.1 - 14 - 15 1725 72
U13897.1 - 14 - 15 1725 72

View Full Product Details