Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLG1
Entrez Gene ID
1739
Gene Name
discs large MAGUK scaffold protein 1
Gene Aliases
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
Chromosome Location
Chr.3: 197042560 - 197299272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098424.1 NP_001091894.1 17 - 18 2126 66
NM_001204386.1 NP_001191315.1 16 - 17 2027 66
NM_001204387.1 NP_001191316.1 12 - 13 1911 66
NM_001204388.1 NP_001191317.1 12 - 13 1911 66
NM_001290983.1 NP_001277912.1 17 - 18 2195 66
NM_004087.2 NP_004078.2 17 - 18 2126 66
XM_005269289.3 XP_005269346.1 17 - 18 2408 66
XM_011512502.2 XP_011510804.1 17 - 18 2408 66
XM_011512503.1 XP_011510805.1 16 - 17 2306 66
XM_011512505.1 XP_011510807.1 16 - 17 2306 66
XM_011512506.1 XP_011510808.1 15 - 16 2252 66
XM_017005800.1 XP_016861289.1 17 - 18 2543 66
XM_017005801.1 XP_016861290.1 17 - 18 2063 66
XM_017005802.1 XP_016861291.1 17 - 18 2133 66
XM_017005803.1 XP_016861292.1 17 - 18 2054 66
XM_017005804.1 XP_016861293.1 17 - 18 2408 66
XM_017005805.1 XP_016861294.1 17 - 18 2225 66
XM_017005806.1 XP_016861295.1 16 - 17 2306 66
XM_017005807.1 XP_016861296.1 16 - 17 2405 66
XM_017005808.1 XP_016861297.1 16 - 17 1964 66
XM_017005809.1 XP_016861298.1 16 - 17 2066 66
XM_017005810.1 XP_016861299.1 16 - 17 1964 66
XM_017005811.1 XP_016861300.1 15 - 16 2252 66
XM_017005812.1 XP_016861301.1 15 - 16 1906 66
XM_017005813.1 XP_016861302.1 15 - 16 1910 66
XM_017005814.1 XP_016861303.1 15 - 16 2013 66
XM_017005815.1 XP_016861304.1 15 - 16 2252 66
XM_017005816.1 XP_016861305.1 16 - 17 2405 66
XM_017005817.1 XP_016861306.1 16 - 17 1936 66
XM_017005818.1 XP_016861307.1 16 - 17 2038 66
XM_017005819.1 XP_016861308.1 16 - 17 1955 66
XM_017005820.1 XP_016861309.1 15 - 16 1970 66
XM_017005821.1 XP_016861310.1 13 - 14 4411 66
XM_017005822.1 XP_016861311.1 13 - 14 4413 66
XM_017005823.1 XP_016861312.1 12 - 13 4308 66
XM_017005824.1 XP_016861313.1 7 - 8 927 66
GenBank mRNA AB209536.2 - 9 - 10 1249 66
AK293967.1 - 12 - 13 1630 66
AK294772.1 - 12 - 13 1638 66
AK294855.1 - 12 - 13 1911 66
AK310596.1 - 16 - 17 1965 66
BC140841.1 - 17 - 18 2091 66
BC144651.1 - 16 - 17 1992 66
EF553524.1 - 17 - 18 2195 66
U13896.1 - 17 - 18 2124 66
U13897.1 - 17 - 18 2124 66

View Full Product Details