Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLG1
Entrez Gene ID
1739
Gene Name
discs large MAGUK scaffold protein 1
Gene Aliases
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
Chromosome Location
Chr.3: 197042560 - 197299272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098424.1 NP_001091894.1 9 - 10 1001 125
NM_001204386.1 NP_001191315.1 8 - 9 902 125
NM_001204387.1 NP_001191316.1 4 - 5 786 125
NM_001204388.1 NP_001191317.1 4 - 5 786 125
NM_001290983.1 NP_001277912.1 9 - 10 1070 125
NM_004087.2 NP_004078.2 9 - 10 1001 125
XM_005269289.3 XP_005269346.1 9 - 10 1283 125
XM_011512502.2 XP_011510804.1 9 - 10 1283 125
XM_011512503.1 XP_011510805.1 8 - 9 1181 125
XM_011512505.1 XP_011510807.1 8 - 9 1181 125
XM_011512506.1 XP_011510808.1 7 - 8 1127 125
XM_011512509.1 XP_011510811.1 8 - 9 1181 125
XM_017005800.1 XP_016861289.1 9 - 10 1418 125
XM_017005801.1 XP_016861290.1 9 - 10 938 125
XM_017005802.1 XP_016861291.1 9 - 10 1008 125
XM_017005803.1 XP_016861292.1 9 - 10 929 125
XM_017005804.1 XP_016861293.1 9 - 10 1283 125
XM_017005805.1 XP_016861294.1 9 - 10 1100 125
XM_017005806.1 XP_016861295.1 8 - 9 1181 125
XM_017005807.1 XP_016861296.1 8 - 9 1280 125
XM_017005808.1 XP_016861297.1 8 - 9 839 125
XM_017005809.1 XP_016861298.1 8 - 9 941 125
XM_017005810.1 XP_016861299.1 8 - 9 839 125
XM_017005811.1 XP_016861300.1 7 - 8 1127 125
XM_017005812.1 XP_016861301.1 7 - 8 781 125
XM_017005813.1 XP_016861302.1 7 - 8 785 125
XM_017005814.1 XP_016861303.1 7 - 8 888 125
XM_017005815.1 XP_016861304.1 7 - 8 1127 125
XM_017005816.1 XP_016861305.1 8 - 9 1280 125
XM_017005817.1 XP_016861306.1 8 - 9 811 125
XM_017005818.1 XP_016861307.1 8 - 9 913 125
XM_017005819.1 XP_016861308.1 8 - 9 830 125
XM_017005820.1 XP_016861309.1 7 - 8 845 125
XM_017005821.1 XP_016861310.1 5 - 6 3286 125
XM_017005822.1 XP_016861311.1 5 - 6 3288 125
XM_017005823.1 XP_016861312.1 4 - 5 3183 125
GenBank mRNA AB209536.2 - 1 - 2 124 125
AK293967.1 - 4 - 5 505 125
AK294772.1 - 4 - 5 513 125
AK294855.1 - 4 - 5 786 125
AK304280.1 - 7 - 8 848 125
AK310596.1 - 8 - 9 840 125
AK310893.1 - 9 - 10 1285 125
AK311129.1 - 3 - 4 238 125
BC015560.2 - 9 - 10 1240 125
BC028486.1 - 9 - 10 1240 125
BC140841.1 - 9 - 10 966 125
BC144651.1 - 8 - 9 867 125
EF553524.1 - 9 - 10 1070 125
U13896.1 - 9 - 10 999 125
U13897.1 - 9 - 10 999 125

View Full Product Details