The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323515.1 | NP_001310444.1 | 18 - 19 | 1817 | 71 |
NM_001323516.1 | NP_001310445.1 | 18 - 19 | 1890 | 71 | |
NM_001323519.1 | NP_001310448.1 | 18 - 19 | 1936 | 71 | |
NM_001323521.1 | NP_001310450.1 | 18 - 19 | 1881 | 71 | |
NM_001323522.1 | NP_001310451.1 | 18 - 19 | 1886 | 71 | |
NM_001323523.1 | NP_001310452.1 | 18 - 19 | 2109 | 71 | |
NM_001323524.1 | NP_001310453.1 | 18 - 19 | 1887 | 71 | |
NM_001323525.1 | NP_001310454.1 | 18 - 19 | 2060 | 71 | |
NM_001323526.1 | NP_001310455.1 | 18 - 19 | 1997 | 71 | |
NM_001323528.1 | NP_001310457.1 | 18 - 19 | 1950 | 71 | |
NM_001323530.1 | NP_001310459.1 | 18 - 19 | 2000 | 71 | |
NM_001323531.1 | NP_001310460.1 | 18 - 19 | 1889 | 71 | |
NM_001323532.1 | NP_001310461.1 | 18 - 19 | 2057 | 71 | |
NM_020214.3 | NP_064599.2 | 18 - 19 | 1947 | 71 | |
NR_136594.1 | 19 - 20 | 2178 | 71 | ||
NR_136596.1 | 18 - 19 | 2200 | 71 | ||
NR_136599.1 | 18 - 19 | 1890 | 71 | ||
NR_136603.1 | 18 - 19 | 1881 | 71 | ||
NR_136604.1 | 19 - 20 | 2118 | 71 | ||
NR_136606.1 | 18 - 19 | 2109 | 71 | ||
NR_136607.1 | 18 - 19 | 2057 | 71 | ||
NR_136608.1 | 18 - 19 | 1947 | 71 | ||
NR_136610.1 | 18 - 19 | 2060 | 71 | ||
NR_136611.1 | 18 - 19 | 1997 | 71 | ||
XM_011521805.1 | XP_011520107.1 | 13 - 14 | 1322 | 71 | |
XM_017022418.1 | XP_016877907.1 | 17 - 18 | 2537 | 71 | |
GenBank mRNA | AB499727.1 | - | 15 - 16 | 1415 | 71 |
AK022547.1 | - | 17 - 18 | 1641 | 71 | |
AK024703.1 | - | 6 - 7 | 485 | 71 | |
AL122091.1 | - | 13 - 14 | 1229 | 71 | |
AL390087.1 | - | 16 - 17 | 1607 | 71 | |
AL390093.1 | - | 5 - 6 | 335 | 71 | |
BC015314.1 | - | 14 - 15 | 1216 | 71 | |
BC026955.1 | - | 14 - 15 | 1212 | 71 | |
BC110902.1 | - | 16 - 17 | 1463 | 71 | |
BC119688.2 | - | 16 - 17 | 1477 | 71 | |
BC119689.1 | - | 16 - 17 | 1477 | 71 |