The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147198.3 | NP_671731.1 | 3 - 4 | 304 | 69 |
GenBank mRNA | AW269752.1 | - | 2 - 3 | 69 | |
AY047610.1 | - | 3 - 4 | 304 | 69 | |
BC069295.1 | - | 2 - 3 | 170 | 69 | |
BC101652.1 | - | 1 - 2 | 139 | 69 | |
BC101656.1 | - | 1 - 2 | 139 | 69 |