Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THEM4
Entrez Gene ID
117145
Gene Name
thioesterase superfamily member 4
Gene Aliases
CTMP
Chromosome Location
Chr.1: 151870866 - 151909885 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053055.4 NP_444283.2 3 - 4 874 75
XM_011509142.2 XP_011507444.1 4 - 5 532 75
GenBank mRNA AJ313515.1 - 3 - 4 493 75
AK096211.1 - 2 - 3 1619 75
AK314852.1 - 3 - 4 447 75
BC065277.1 - 3 - 4 533 75

View Full Product Details