The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005262400.3 | XP_005262457.1 | 9 - 10 | 1357 | 79 |
XM_011531317.2 | XP_011529619.1 | 9 - 10 | 1357 | 79 | |
XM_011531318.2 | XP_011529620.1 | 9 - 10 | 1357 | 79 |