Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWA5A
Entrez Gene ID
4013
Gene Name
von Willebrand factor A domain containing 5A
Gene Aliases
BCSC-1, BCSC1, LOH11CR2A
Chromosome Location
Chr.11: 124115404 - 124146912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130142.1 NP_001123614.1 12 - 13 1566 96
NM_014622.4 NP_055437.2 11 - 12 1451 96
XM_011542828.2 XP_011541130.1 12 - 13 1677 96
GenBank mRNA AF002672.1 - 10 - 11 1363 96
AK291698.1 - 11 - 12 1405 96
AK314934.1 - 4 - 5 416 96
AY366505.1 - 11 - 12 1451 96
AY366508.1 - 12 - 13 1566 96

View Full Product Details