Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA5A
Entrez Gene ID
4013
Gene Name
von Willebrand factor A domain containing 5A
Gene Aliases
BCSC-1, BCSC1, LOH11CR2A
Chromosome Location
Chr.11: 124115404 - 124146912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130142.1 NP_001123614.1 14 - 15 1827 80
NM_014622.4 NP_055437.2 13 - 14 1712 80
XM_011542828.2 XP_011541130.1 14 - 15 1938 80
GenBank mRNA AF002672.1 - 12 - 13 1624 80
AK291698.1 - 13 - 14 1666 80
AK314934.1 - 6 - 7 677 80
AY366505.1 - 13 - 14 1712 80
AY366508.1 - 14 - 15 1827 80

View Full Product Details