Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA5A
Entrez Gene ID
4013
Gene Name
von Willebrand factor A domain containing 5A
Gene Aliases
BCSC-1, BCSC1, LOH11CR2A
Chromosome Location
Chr.11: 124115404 - 124146912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130142.1 NP_001123614.1 18 - 19 2488 90
NM_014622.4 NP_055437.2 17 - 18 2373 90
XM_011542828.2 XP_011541130.1 18 - 19 2599 90
GenBank mRNA AF002672.1 - 16 - 17 2285 90
AK291698.1 - 17 - 18 2327 90
AK314934.1 - 10 - 11 1338 90
AY366504.1 - 8 - 9 950 90
AY366505.1 - 17 - 18 2373 90
AY366507.1 - 9 - 10 1065 90
AY366508.1 - 18 - 19 2488 90

View Full Product Details