Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD15
Entrez Gene ID
79047
Gene Name
potassium channel tetramerization domain containing 15
Gene Aliases
-
Chromosome Location
Chr.19: 33795540 - 33815761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129994.1 NP_001123466.1 5 - 6 646 85
NM_001129995.1 NP_001123467.1 5 - 6 643 85
NM_024076.2 NP_076981.2 5 - 6 646 85
XM_011527296.1 XP_011525598.1 5 - 6 629 85
XM_011527297.2 XP_011525599.1 6 - 7 680 85
XM_011527298.2 XP_011525600.1 6 - 7 715 85
XM_017027283.1 XP_016882772.1 6 - 7 771 85
GenBank mRNA AK027901.1 - 5 - 6 639 85
AK096843.1 - 4 - 5 668 85
AK225793.1 - 5 - 6 611 85
AK291465.1 - 5 - 6 631 85
AK294543.1 - 6 - 7 715 85
BC001185.1 - 5 - 6 630 85
BC009335.2 - 5 - 6 630 85
CN301512.1 - 5 - 6 653 85

View Full Product Details