Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMK2D
Entrez Gene ID
817
Gene Name
calcium/calmodulin dependent protein kinase II delta
Gene Aliases
CAMKD
Chromosome Location
Chr.4: 113451032 - 113761927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001221.3 NP_001212.2 18 - 19 2385 72
NM_001321566.1 NP_001308495.1 19 - 20 2427 72
NM_001321569.1 NP_001308498.1 20 - 21 2487 72
NM_001321571.1 NP_001308500.1 20 - 21 2487 72
NM_001321573.1 NP_001308502.1 20 - 21 2460 72
NM_001321574.1 NP_001308503.1 18 - 19 2382 72
NM_001321580.1 NP_001308509.1 19 - 20 2445 72
NM_001321588.1 NP_001308517.1 19 - 20 2424 72
XM_005263253.3 XP_005263310.1 20 - 21 2302 72
XM_005263254.3 XP_005263311.1 20 - 21 2323 72
XM_011532289.1 XP_011530591.1 19 - 20 2236 72
XM_011532290.1 XP_011530592.1 19 - 20 2233 72
XM_011532292.2 XP_011530594.1 16 - 17 1337 72
XM_011532295.1 XP_011530597.1 19 - 20 2257 72
XM_017008672.1 XP_016864161.1 19 - 20 2266 72
XM_017008673.1 XP_016864162.1 19 - 20 2263 72
XM_017008675.1 XP_016864164.1 21 - 22 2427 72
XM_017008676.1 XP_016864165.1 20 - 21 1663 72
GenBank mRNA AK294517.1 - 20 - 21 1778 72
AL120285.1 - 3 - 4 338 72
CF551834.1 - 2 - 3 186 72

View Full Product Details