The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271686.1 | NP_001258615.1 | 1 - 2 | 230 | 107 |
XM_005274140.4 | XP_005274197.1 | 1 - 2 | 219 | 107 | |
XM_006718647.3 | XP_006718710.1 | 1 - 2 | 221 | 107 | |
XM_011545196.2 | XP_011543498.1 | 1 - 2 | 225 | 107 | |
XM_011545199.2 | XP_011543501.1 | 1 - 2 | 227 | 107 | |
XM_011545201.2 | XP_011543503.1 | 1 - 2 | 232 | 107 | |
GenBank mRNA | BC051820.1 | - | 1 - 2 | 230 | 107 |