Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DENND1A
Entrez Gene ID
57706
Gene Name
DENN domain containing 1A
Gene Aliases
FAM31A, KIAA1608
Chromosome Location
Chr.9: 123379654 - 123930158 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020946.1 NP_065997.1 6 - 7 603 75
NM_024820.2 NP_079096.2 6 - 7 603 75
XM_005252109.2 XP_005252166.1 6 - 7 452 75
XM_005252111.4 XP_005252168.1 6 - 7 1419 75
XM_005252113.4 XP_005252170.1 6 - 7 1408 75
XM_006717195.3 XP_006717258.1 6 - 7 1420 75
XM_011518882.2 XP_011517184.1 6 - 7 1422 75
XM_011518883.2 XP_011517185.1 6 - 7 1423 75
XM_011518884.2 XP_011517186.1 5 - 6 1373 75
XM_011518885.2 XP_011517187.1 5 - 6 1115 75
XM_011518886.2 XP_011517188.1 6 - 7 1420 75
XM_011518887.2 XP_011517189.1 6 - 7 1411 75
XM_017014947.1 XP_016870436.1 5 - 6 1378 75
XM_017014948.1 XP_016870437.1 6 - 7 452 75
XM_017014949.1 XP_016870438.1 6 - 7 1423 75
XM_017014950.1 XP_016870439.1 6 - 7 1422 75
XM_017014951.1 XP_016870440.1 5 - 6 396 75
XM_017014952.1 XP_016870441.1 5 - 6 579 75
GenBank mRNA AK024782.1 - 6 - 7 591 75
AK225104.1 - 6 - 7 591 75
AK295710.1 - 4 - 5 464 75
BC009616.2 - 5 - 6 571 75
BC028061.1 - 5 - 6 393 75
BC039703.1 - 6 - 7 456 75

View Full Product Details