The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146312.2 | NP_001139784.1 | 6 - 7 | 892 | 78 |
NM_153604.3 | NP_705832.1 | 6 - 7 | 892 | 78 | |
XM_005256863.1 | XP_005256920.1 | 6 - 7 | 892 | 78 | |
XM_005256864.1 | XP_005256921.1 | 7 - 8 | 936 | 78 | |
GenBank mRNA | AF532596.1 | - | 6 - 7 | 593 | 78 |
AK097821.1 | - | 3 - 4 | 514 | 78 | |
AK128608.1 | - | 6 - 7 | 792 | 78 | |
AK292885.1 | - | 6 - 7 | 761 | 78 | |
AY764180.1 | - | 6 - 7 | 885 | 78 | |
BC126307.1 | - | 6 - 7 | 651 | 78 | |
BC143391.1 | - | 6 - 7 | 651 | 78 |