Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOCD
Entrez Gene ID
93649
Gene Name
myocardin
Gene Aliases
MYCD
Chromosome Location
Chr.17: 12665890 - 12768949 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146312.2 NP_001139784.1 9 - 10 1422 65
NM_153604.3 NP_705832.1 9 - 10 1422 65
XM_005256863.1 XP_005256920.1 9 - 10 1422 65
XM_005256864.1 XP_005256921.1 10 - 11 1466 65
XM_017025342.1 XP_016880831.1 8 - 9 1296 65
GenBank mRNA AF532596.1 - 9 - 10 1123 65
AK097821.1 - 6 - 7 1044 65
AK292885.1 - 9 - 10 1291 65
AY764180.1 - 9 - 10 1415 65
BC126307.1 - 9 - 10 1181 65
BC143391.1 - 9 - 10 1181 65
BX640673.1 - 2 - 3 238 65

View Full Product Details