Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM53C
Entrez Gene ID
51307
Gene Name
family with sequence similarity 53 member C
Gene Aliases
C5orf6
Chromosome Location
Chr.5: 138337535 - 138349729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016605.2 NP_057689.1 1 - 2 291 133

View Full Product Details