Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM53C
Entrez Gene ID
51307
Gene Name
family with sequence similarity 53 member C
Gene Aliases
C5orf6
Chromosome Location
Chr.5: 138337535 - 138349729 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135647.1 NP_001129119.1 4 - 5 1384 74
NM_016605.2 NP_057689.1 4 - 5 1359 74
GenBank mRNA AF251040.1 - 4 - 5 1095 74
AK027412.1 - 2 - 3 250 74
AK310378.1 - 4 - 5 537 74
AK315567.1 - 4 - 5 1108 74
BC000259.1 - 4 - 5 1101 74
BC052993.1 - 4 - 5 1324 74

View Full Product Details