The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025014.1 | NP_079290.1 | 4 - 5 | 1084 | 76 |
NM_173728.3 | NP_776089.2 | 4 - 5 | 1148 | 76 | |
XM_011523734.1 | XP_011522036.1 | 4 - 5 | 1148 | 76 | |
XM_011523735.1 | XP_011522037.1 | 4 - 5 | 1084 | 76 | |
XM_011523736.1 | XP_011522038.1 | 4 - 5 | 1148 | 76 | |
GenBank mRNA | AB020722.1 | - | 4 - 5 | 1064 | 76 |
AK291626.1 | - | 4 - 5 | 1108 | 76 | |
AK300327.1 | - | 4 - 5 | 569 | 76 | |
BC036749.1 | - | 4 - 5 | 1103 | 76 | |
KU178557.1 | - | 3 - 4 | 993 | 76 | |
KU178558.1 | - | 3 - 4 | 993 | 76 |