The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025014.1 | NP_079290.1 | 10 - 11 | 1871 | 112 |
NM_173728.3 | NP_776089.2 | 10 - 11 | 1935 | 112 | |
XM_011523734.1 | XP_011522036.1 | 10 - 11 | 1980 | 112 | |
XM_011523735.1 | XP_011522037.1 | 10 - 11 | 1916 | 112 | |
XM_011523736.1 | XP_011522038.1 | 10 - 11 | 1980 | 112 | |
XM_011523737.1 | XP_011522039.1 | 7 - 8 | 972 | 112 | |
GenBank mRNA | AB020722.1 | - | 10 - 11 | 1851 | 112 |
AK291626.1 | - | 10 - 11 | 1895 | 112 | |
AK300327.1 | - | 10 - 11 | 1356 | 112 | |
BC036749.1 | - | 10 - 11 | 1890 | 112 | |
KU178557.1 | - | 9 - 10 | 1780 | 112 | |
KU178558.1 | - | 9 - 10 | 1780 | 112 |