Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TEPSIN
Entrez Gene ID
146705
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
Gene Aliases
C17orf56, ENTHD2
Chromosome Location
Chr.17: 81228277 - 81239092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144679.2 NP_653280.1 3 - 4 271 97
XM_005257066.2 XP_005257123.1 4 - 5 865 97
XM_005257067.2 XP_005257124.1 3 - 4 271 97
XM_006721709.3 XP_006721772.1 3 - 4 278 97
XM_006721712.2 XP_006721775.1 3 - 4 709 97
XM_011524355.1 XP_011522657.1 4 - 5 871 97
XM_011524356.1 XP_011522658.1 4 - 5 871 97
XM_011524357.1 XP_011522659.1 3 - 4 899 97
XM_011524358.2 XP_011522660.1 4 - 5 800 97
XM_017024202.1 XP_016879691.1 4 - 5 1112 97
XM_017024203.1 XP_016879692.1 5 - 6 759 97
XM_017024204.1 XP_016879693.1 4 - 5 1312 97
GenBank mRNA AK056090.1 - 3 - 4 235 97
AK127221.1 - 1 - 2 1424 97
AK128728.1 - 4 - 5 1584 97
BC064483.1 - 3 - 4 232 97
DB122720.1 - 3 - 4 271 97

View Full Product Details