Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEPSIN
Entrez Gene ID
146705
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
Gene Aliases
C17orf56, ENTHD2
Chromosome Location
Chr.17: 81228277 - 81239092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144679.2 NP_653280.1 1 - 2 108 56
XM_005257067.2 XP_005257124.1 1 - 2 108 56
XM_006721709.3 XP_006721772.1 1 - 2 109 56
GenBank mRNA AK056090.1 - 1 - 2 72 56
BC064483.1 - 1 - 2 69 56
DB122720.1 - 1 - 2 108 56

View Full Product Details