The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321567.1 | NP_001308496.1 | 19 - 20 | 2439 | 64 |
NM_001321579.1 | NP_001308508.1 | 19 - 20 | 2439 | 64 | |
NM_001321583.1 | NP_001308512.1 | 18 - 19 | 2379 | 64 | |
NM_001321589.1 | NP_001308518.1 | 18 - 19 | 2379 | 64 | |
NM_001321591.1 | NP_001308520.1 | 19 - 20 | 2412 | 64 | |
NM_172127.2 | NP_742125.1 | 17 - 18 | 2337 | 64 | |
XM_006714330.2 | XP_006714393.1 | 17 - 18 | 2176 | 64 | |
XM_011532291.1 | XP_011530593.1 | 18 - 19 | 2188 | 64 | |
XM_017008677.1 | XP_016864166.1 | 18 - 19 | 2215 | 64 | |
XM_017008678.1 | XP_016864167.1 | 17 - 18 | 2152 | 64 | |
GenBank mRNA | AK025368.1 | - | 6 - 7 | 588 | 64 |