The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000450.2 | NP_000441.2 | 3 - 4 | 575 | 141 |
GenBank mRNA | BC131551.1 | - | 3 - 4 | 534 | 141 |
BC142677.1 | - | 3 - 4 | 562 | 141 | |
BC142711.1 | - | 3 - 4 | 562 | 141 | |
M24736.1 | - | 3 - 4 | 534 | 141 | |
M30640.1 | - | 3 - 4 | 558 | 141 |