Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SELE
Entrez Gene ID
6401
Gene Name
selectin E
Gene Aliases
CD62E, ELAM, ELAM1, ESEL, LECAM2
Chromosome Location
Chr.1: 169722640 - 169734079 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000450.2 NP_000441.2 8 - 9 1437 152
GenBank mRNA BC131551.1 - 8 - 9 1396 152
BC142677.1 - 8 - 9 1424 152
BC142711.1 - 8 - 9 1424 152
M24736.1 - 8 - 9 1396 152
M30640.1 - 8 - 9 1420 152

View Full Product Details