Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADD1
Entrez Gene ID
118
Gene Name
adducin 1
Gene Aliases
ADDA
Chromosome Location
Chr.4: 2843732 - 2930076 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001119.4 NP_001110.2 4 - 5 692 65
NM_001286645.1 NP_001273574.1 5 - 6 1367 65
NM_014189.3 NP_054908.2 4 - 5 692 65
NM_014190.3 NP_054909.2 4 - 5 692 65
NM_176801.2 NP_789771.1 4 - 5 692 65
XM_005247933.2 XP_005247990.1 4 - 5 817 65
XM_005247934.2 XP_005247991.1 4 - 5 781 65
XM_005247935.2 XP_005247992.1 4 - 5 817 65
XM_005247936.2 XP_005247993.1 4 - 5 817 65
XM_005247937.2 XP_005247994.1 4 - 5 817 65
XM_017007703.1 XP_016863192.1 4 - 5 542 65
XM_017007704.1 XP_016863193.1 4 - 5 781 65
XM_017007705.1 XP_016863194.1 4 - 5 781 65
XM_017007706.1 XP_016863195.1 4 - 5 781 65
XM_017007707.1 XP_016863196.1 3 - 4 539 65
XM_017007708.1 XP_016863197.1 4 - 5 817 65
XM_017007709.1 XP_016863198.1 4 - 5 781 65
XM_017007710.1 XP_016863199.1 3 - 4 539 65
GenBank mRNA AK025413.1 - 2 - 3 297 65
AK295457.1 - 4 - 5 779 65
AK309186.1 - 4 - 5 570 65
BC013393.2 - 4 - 5 627 65
BC042998.1 - 5 - 6 1372 65
CR995783.1 - 4 - 5 667 65
D44632.1 - 4 - 5 709 65
L07261.1 - 3 - 4 506 65

View Full Product Details