Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEK
Entrez Gene ID
5341
Gene Name
pleckstrin
Gene Aliases
P47
Chromosome Location
Chr.2: 68365190 - 68397453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002664.2 NP_002655.2 6 - 7 925 95
GenBank mRNA AB208967.1 - 6 - 7 819 95
AK313756.1 - 6 - 7 847 95
BC018549.1 - 6 - 7 831 95
CR542056.1 - 6 - 7 763 95
CR542079.1 - 6 - 7 763 95
X07743.1 - 6 - 7 823 95

View Full Product Details