Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IGF1R
Entrez Gene ID
3480
Gene Name
insulin like growth factor 1 receptor
Gene Aliases
CD221, IGFIR, IGFR, JTK13
Chromosome Location
Chr.15: 98648539 - 98964530 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000875.4 NP_000866.1 9 - 10 3043 64
NM_001291858.1 NP_001278787.1 9 - 10 3043 64
XM_011521516.2 XP_011519818.1 8 - 9 1493 64
XM_011521517.2 XP_011519819.1 5 - 6 906 64
XM_017022136.1 XP_016877625.1 9 - 10 2546 64
XM_017022137.1 XP_016877626.1 9 - 10 2546 64
XM_017022138.1 XP_016877627.1 9 - 10 2546 64
XM_017022139.1 XP_016877628.1 9 - 10 1984 64
GenBank mRNA AB425196.1 - 9 - 10 2529 64
BC113610.1 - 9 - 10 2032 64
BC113612.1 - 9 - 10 2032 64
BC143721.1 - 9 - 10 2032 64
EU826611.1 - 9 - 10 2000 64
EU826612.1 - 9 - 10 2000 64
X04434.1 - 9 - 10 2045 64

View Full Product Details