The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153235.3 | NP_694967.3 | 7 - 8 | 1242 | 76 |
XM_005266836.1 | XP_005266893.1 | 8 - 9 | 1609 | 76 | |
XM_011535505.2 | XP_011533807.1 | 7 - 8 | 1346 | 76 | |
XM_011535506.2 | XP_011533808.1 | 6 - 7 | 792 | 76 | |
XM_017010322.1 | XP_016865811.1 | 8 - 9 | 1472 | 76 | |
GenBank mRNA | AB085905.1 | - | 7 - 8 | 1242 | 76 |
AK314725.1 | - | 7 - 8 | 1190 | 76 | |
AL831988.1 | - | 7 - 8 | 1180 | 76 | |
AL832322.1 | - | 4 - 5 | 540 | 76 | |
AL834248.2 | - | 7 - 8 | 1170 | 76 | |
BC115383.1 | - | 7 - 8 | 1197 | 76 | |
BC115384.2 | - | 7 - 8 | 1197 | 76 | |
BX647477.1 | - | 7 - 8 | 1190 | 76 | |
BX647633.1 | - | 7 - 8 | 1168 | 76 |