Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCCHC8
Entrez Gene ID
55596
Gene Name
zinc finger CCHC-type containing 8
Gene Aliases
-
Chromosome Location
Chr.12: 122471599 - 122501073 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017612.4 NP_060082.2 11 - 12 1372 98
XM_006719491.2 XP_006719554.1 8 - 9 1057 98
XM_011538555.2 XP_011536857.1 10 - 11 1914 98
XM_017019606.1 XP_016875095.1 11 - 12 1292 98
GenBank mRNA AK027702.1 - 11 - 12 1241 98
AK074638.1 - 11 - 12 1270 98
AL157433.1 - 9 - 10 2781 98
BC017704.2 - 4 - 5 496 98
BC065918.1 - 11 - 12 1211 98

View Full Product Details