Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCCHC8
Entrez Gene ID
55596
Gene Name
zinc finger CCHC-type containing 8
Gene Aliases
-
Chromosome Location
Chr.12: 122471599 - 122501073 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017612.4 NP_060082.2 7 - 8 901 115
XM_011538555.2 XP_011536857.1 6 - 7 1443 115
XM_017019606.1 XP_016875095.1 7 - 8 821 115
GenBank mRNA AK027702.1 - 7 - 8 770 115
AK074638.1 - 7 - 8 799 115
AK291174.1 - 3 - 4 236 115
AL157433.1 - 5 - 6 2310 115
BC065918.1 - 7 - 8 740 115

View Full Product Details