The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007050.5 | NP_008981.4 | 16 - 17 | 2676 | 92 |
NM_133170.3 | NP_573400.3 | 17 - 18 | 2733 | 92 | |
XM_017027611.1 | XP_016883100.1 | 15 - 16 | 2709 | 92 | |
XM_017027612.1 | XP_016883101.1 | 16 - 17 | 2766 | 92 | |
XM_017027613.1 | XP_016883102.1 | 15 - 16 | 2708 | 92 | |
GenBank mRNA | AB006621.2 | - | 16 - 17 | 2584 | 92 |
AF043644.4 | - | 17 - 18 | 2742 | 92 | |
BC153300.1 | - | 16 - 17 | 2584 | 92 |