The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007050.5 | NP_008981.4 | 20 - 21 | 3070 | 75 |
NM_133170.3 | NP_573400.3 | 21 - 22 | 3127 | 75 | |
XM_017027611.1 | XP_016883100.1 | 19 - 20 | 3103 | 75 | |
XM_017027612.1 | XP_016883101.1 | 20 - 21 | 3160 | 75 | |
XM_017027613.1 | XP_016883102.1 | 19 - 20 | 3102 | 75 | |
GenBank mRNA | AB006621.2 | - | 20 - 21 | 2978 | 75 |
AF043644.4 | - | 21 - 22 | 3136 | 75 | |
BC153300.1 | - | 20 - 21 | 2978 | 75 |