The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007050.5 | NP_008981.4 | 29 - 30 | 4315 | 68 |
NM_133170.3 | NP_573400.3 | 30 - 31 | 4372 | 68 | |
XM_017027611.1 | XP_016883100.1 | 29 - 30 | 4408 | 68 | |
XM_017027612.1 | XP_016883101.1 | 29 - 30 | 4405 | 68 | |
XM_017027613.1 | XP_016883102.1 | 28 - 29 | 4347 | 68 | |
GenBank mRNA | AB006621.2 | - | 29 - 30 | 4223 | 68 |
AF043644.4 | - | 30 - 31 | 4381 | 68 | |
BC153300.1 | - | 29 - 30 | 4223 | 68 |