The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271629.1 | NP_001258558.1 | 9 - 10 | 1443 | 110 |
NM_005920.3 | NP_005911.1 | 10 - 11 | 1745 | 110 | |
XM_005245169.4 | XP_005245226.1 | 10 - 11 | 1727 | 110 | |
XM_005245170.3 | XP_005245227.1 | 10 - 11 | 1752 | 110 | |
XM_006711330.3 | XP_006711393.1 | 10 - 11 | 1456 | 110 | |
XM_006711332.3 | XP_006711395.1 | 10 - 11 | 1740 | 110 | |
XM_006711333.2 | XP_006711396.1 | 9 - 10 | 1719 | 110 | |
XM_006711334.3 | XP_006711397.1 | 9 - 10 | 1717 | 110 | |
XM_011509569.2 | XP_011507871.1 | 10 - 11 | 1777 | 110 | |
XM_017001314.1 | XP_016856803.1 | 9 - 10 | 1861 | 110 | |
XM_017001315.1 | XP_016856804.1 | 9 - 10 | 1456 | 110 | |
GenBank mRNA | AB210005.1 | - | 10 - 11 | 1659 | 110 |
AK308641.1 | - | 10 - 11 | 1645 | 110 | |
BC032479.1 | - | 7 - 8 | 1294 | 110 | |
BC040949.1 | - | 9 - 10 | 1443 | 110 | |
BC054520.1 | - | 10 - 11 | 1640 | 110 | |
L16794.1 | - | 10 - 11 | 1445 | 110 |