The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321702.1 | NP_001308631.1 | 1 - 2 | 72 | 61 |
GenBank mRNA | BM313608.1 | - | 1 - 2 | 72 | 61 |