The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002836.3 | NP_002827.1 | 6 - 7 | 620 | 112 |
NM_080840.2 | NP_543030.1 | 2 - 3 | 338 | 112 | |
NM_080841.2 | NP_543031.1 | 1 - 2 | 73 | 202 | |
GenBank mRNA | AB209702.1 | - | 2 - 3 | 284 | 112 |
AK290233.1 | - | 2 - 3 | 277 | 112 | |
AK294990.1 | - | 2 - 3 | 298 | 112 | |
BC027308.1 | - | 2 - 3 | 143 | 112 | |
BI597187.1 | - | 2 - 3 | 301 | 112 | |
BQ213974.1 | - | 2 - 3 | 338 | 112 | |
BX571753.1 | - | 3 - 4 | 389 | 112 | |
M34668.1 | - | 6 - 7 | 640 | 112 | |
X54890.1 | - | 2 - 3 | 131 | 112 |