Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KPNA3
Entrez Gene ID
3839
Gene Name
karyopherin subunit alpha 3
Gene Aliases
IPOA4, SRP1, SRP1gamma, SRP4, hSRP1
Chromosome Location
Chr.13: 49699307 - 49792921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002267.3 NP_002258.2 5 - 6 704 78
XM_017020561.1 XP_016876050.1 5 - 6 387 78
GenBank mRNA AF005263.1 - 5 - 6 301 78
AF034756.1 - 5 - 6 291 78
AK223480.1 - 5 - 6 326 78
AK290528.1 - 5 - 6 364 78
AK291000.1 - 5 - 6 328 78
BC017355.2 - 5 - 6 313 78
BC024202.2 - 5 - 6 313 78
BC035090.2 - 5 - 6 705 78
DN991804.1 - 5 - 6 492 78
Y12394.1 - 5 - 6 304 78

View Full Product Details