The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166102.1 | NP_001159574.1 | 8 - 9 | 1288 | 66 |
NM_007103.3 | NP_009034.2 | 8 - 9 | 1315 | 66 | |
GenBank mRNA | AF053070.1 | - | 8 - 9 | 1231 | 66 |
AF092131.1 | - | 8 - 9 | 1241 | 66 | |
AK055875.1 | - | 5 - 6 | 2394 | 66 | |
AK223061.1 | - | 8 - 9 | 1209 | 66 | |
AK293522.1 | - | 7 - 8 | 1126 | 66 | |
AK300726.1 | - | 6 - 7 | 822 | 66 | |
AK316122.1 | - | 8 - 9 | 1156 | 66 | |
BC007619.1 | - | 5 - 6 | 832 | 66 | |
BC008146.1 | - | 8 - 9 | 1200 | 66 | |
BC015645.2 | - | 8 - 9 | 1195 | 66 | |
CR456739.1 | - | 8 - 9 | 1162 | 66 |