The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126328.2 | NP_001119800.1 | 7 - 8 | 1776 | 146 |
NM_032622.2 | NP_116011.2 | 6 - 7 | 1507 | 146 | |
XM_005265785.4 | XP_005265842.1 | 7 - 8 | 1934 | 146 | |
XM_017008776.1 | XP_016864265.1 | 8 - 9 | 1995 | 146 | |
GenBank mRNA | AF237782.1 | - | 6 - 7 | 1438 | 146 |
AK056823.1 | - | 7 - 8 | 1749 | 146 | |
AK294611.1 | - | 7 - 8 | 1305 | 146 | |
AY358326.1 | - | 6 - 7 | 1418 | 146 | |
BC022983.1 | - | 7 - 8 | 1652 | 146 | |
BC034737.1 | - | 6 - 7 | 1427 | 146 | |
BC094772.1 | - | 3 - 4 | 955 | 146 |